Alfa Aesar™ 2-(Methylthio)naphthalene, 98%

Manufacturer: Alfa Aesar™ B2236803

Quantity 1g

 View more versions of this product

Catalog No. AAB2236803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7433-79-6
    Molecular Formula C11H10S
    Formula Weight 174.27
    MDL Number MFCD00051615
    Beilstein 1859409
    Synonym 2-(Methylmercapto)naphthalene
    Chemical Name or Material 2-(Methylthio)naphthalene

    Specifications

    Formula Weight 174.27
    Molecular Formula C11H10S
    MDL Number MFCD00051615
    CAS 7433-79-6
    Beilstein 1859409
    Synonym 2-(Methylmercapto)naphthalene
    Melting Point 62° to 63°C
    Quantity 1g
    Odor Stench
    Chemical Name or Material 2-(Methylthio)naphthalene
    Assay 98%

    Description

    Applications
    2-(Methylthio)naphthalene is used to produce 1-chloro-2-methylsulfanylnaphthalene. This reaction will need reagent CuCl2-Al2O3, and the menstruum benzene

    Solubility
    Slightly soluble in water

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit