Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-(Methylthio)oxazolo[5,4-c]pyridine, Thermo Scientific™
Click to view available options
Quantity:
1g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
CAS | 169205-96-3 |
Melting Point | 79°C to 81°C |
Molecular Formula | C7H6N2OS |
MDL Number | MFCD08447111 |
Quantity | 1g |
Synonym | 2-methylthio oxazolo 5,4-c pyridine, 2-methylsulfanyl-oxazolo 5,4-c pyridine, acmc-1bzgj, 2-methylsulfanyloxazolo 5,4-c pyridine, 2-methylsulfanyl 1,3 oxazolo 5,4-c pyridine, 2-methylsulfanyl pyrido 4,3-d 1,3 oxazole, 2-methylsulfanyl-1,3 oxazolo 5,4-c pyridine |
InChI Key | DEGNWJBCNBPOPI-UHFFFAOYSA-N |
SMILES | CSC1=NC2=CC=NC=C2O1 |
IUPAC Name | 2-methylsulfanyl-[1,3]oxazolo[5,4-c]pyridine |
Molecular Weight (g/mol) | 166.20 |
Show More |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures