Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Methylthio)pyrazine, 99%

Click to view available options
Quantity:
1g
5g
21948-70-9
C5H6N2S
126.177
MFCD00041024
KBPBOWBQRUXMFV-UHFFFAOYSA-N
2-methylthio pyrazine,2-methylthiopyrazine,pyrazine, methylthio,pyrazinyl methyl sulfide,2-methylsulfanyl pyrazine,methyl pyrazinyl sulfide,methylthio pyrazine,2-methylsulfanyl-pyrazine,pyrazine, 2-methylthio,unii-r1c97e3a4f
519907
2-methylsulfanylpyrazine
CSC1=NC=CN=C1
21948-70-9
C5H6N2S
126.177
MFCD00041024
KBPBOWBQRUXMFV-UHFFFAOYSA-N
2-methylthio pyrazine,2-methylthiopyrazine,pyrazine, methylthio,pyrazinyl methyl sulfide,2-methylsulfanyl pyrazine,methyl pyrazinyl sulfide,methylthio pyrazine,2-methylsulfanyl-pyrazine,pyrazine, 2-methylthio,unii-r1c97e3a4f
519907
2-methylsulfanylpyrazine
CSC1=NC=CN=C1
Melting Point 42°C to 46°C
Quantity 1g
Beilstein 878423
Formula Weight 126.18
Percent Purity 99%
Chemical Name or Material 2-(Methylthio)pyrazine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA