Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Methylthio)thiophene, 97%, Thermo Scientific Chemicals

Catalog Number aaa1176306
Click to view available options
Quantity:
5 g
25 g
5780-36-9
C5H6S2
130.223
MFCD00052382
ZLSMPEVZXWDWEK-UHFFFAOYSA-N
79844
2-methylsulfanylthiophene
CSC1=CC=CS1
5780-36-9
C5H6S2
130.223
MFCD00052382
ZLSMPEVZXWDWEK-UHFFFAOYSA-N
79844
2-methylsulfanylthiophene
CSC1=CC=CS1

2-(Methylthio)thiophene is used as an intermediate in synthetic chemistry. It is also used in the preparation of 2-thiocyanato-thiophene by reaction with cyanogen iodide using sodium methylmercatide as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)thiophene is used as an intermediate in synthetic chemistry. It is also used in the preparation of 2-thiocyanato-thiophene by reaction with cyanogen iodide using sodium methylmercatide as a reagent.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Boiling Point 82°C to 84°C (15 mmHg)
Odor Stench
Refractive Index 1.596
Quantity 5 g
UN Number UN3334
Beilstein 106272
Synonym 2-methylthio thiophene, thiophene, 2-methylthio, 2-methylsulfanyl thiophene, 2-methylthiothiophene, 2-mehtylthio thiophene, thiophene,2-methylthio, acmc-20aomi, 2-methylthio-thiophene, 2-methylmercaptothiophene, 2-methylsulfanyl thiophene #
Formula Weight 130.23
Percent Purity 97%
Chemical Name or Material 2- (Methylthio)thiophene
Hazard Category H319-H335
Hazard Statement GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit