Alfa Aesar™ 2-(Methylthio)thiophene, 97%

Manufacturer: Alfa Aesar™ A1176303

Quantity 1g

 View more versions of this product

Catalog No. AAA1176303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5780-36-9
    Molecular Formula C5H6S2
    Formula Weight 130.23
    MDL Number MFCD00052382
    UN Number UN3334
    Beilstein 106272
    Synonym 2-(Methylmercapto)thiophene; Methyl 2-thienyl sulfide
    Chemical Name or Material 2- (Methylthio)thiophene

    Specifications

    Formula Weight 130.23
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 5780-36-9
    Synonym 2-(Methylmercapto)thiophene; Methyl 2-thienyl sulfide
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
    Molecular Formula C5H6S2
    MDL Number MFCD00052382
    UN Number UN3334
    Beilstein 106272
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H319-H335
    Causes serious eye irritation.
    May cause respiratory irritation.
    Odor Stench
    Quantity 1g
    Boiling Point 82° to 84°C (15mmHg)
    Chemical Name or Material 2- (Methylthio)thiophene
    Assay 97%

    Description

    Applications
    2-(Methylthio)thiophene is used as an intermediate in synthetic chemistry. It is also used in the preparation of 2-thiocyanato-thiophene by reaction with cyanogen iodide using sodium methylmercatide as a reagent.

    Notes
    Incompatible with oxidizing agents.

    GHS H Statement
    H319-H335
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit