Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylthiophene, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
554-14-3
C5H6S
98.16
MFCD00005451
XQQBUAPQHNYYRS-UHFFFAOYSA-N
thiophene, 2-methyl,2-methyl thiophene,thiophene, methyl,alpha-methylthiophene,ccris 2936,2-methyl-thiophene,unii-7115jap77a,2-methylthiacyclopentadiene,a-thiotolene,pubchem5204
11126
2-methylthiophene
CC1=CC=CS1
554-14-3
C5H6S
98.16
MFCD00005451
XQQBUAPQHNYYRS-UHFFFAOYSA-N
thiophene, 2-methyl,2-methyl thiophene,thiophene, methyl,alpha-methylthiophene,ccris 2936,2-methyl-thiophene,unii-7115jap77a,2-methylthiacyclopentadiene,a-thiotolene,pubchem5204
11126
2-methylthiophene
CC1=CC=CS1
Melting Point -63°C
Flash Point 7°C
Boiling Point 113°C
Specific Gravity 1.02
Density 1.0200g/mL
Refractive Index 1.5190 to 1.521
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 17, 37
Solubility Information Solubility in water: immiscible
Formula Weight 98.16
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2-Methylthiophene
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger