Alfa Aesar™ 2-Methylvaleraldehyde, 98%

Manufacturer: Alfa Aesar™ B2419936

Quantity 500g

 View more versions of this product

Catalog No. AAB2419936

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 123-15-9
    Molecular Formula CH3(CH2)2CH(CH3)CHO
    Formula Weight 100.16
    MDL Number MFCD00006986
    UN Number UN2367
    Beilstein 1739423
    Synonym 2-Methylpentanal
    Chemical Name or Material 2-Methylvaleraldehyde

    Specifications

    Formula Weight 100.16
    Molecular Formula CH3(CH2)2CH(CH3)CHO
    UN Number UN2367
    Beilstein 1739423
    Synonym 2-Methylpentanal
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 16°C (62°F)
    Odor Like Aldehyde
    Boiling Point 119° to 120°C
    Melting Point −100°C
    Quantity 500g
    TSCA TSCA
    MDL Number MFCD00006986
    CAS 123-15-9
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALPHA-METHYLVALERALDEHYDE
    RTECS Number RZ6905000
    Chemical Name or Material 2-Methylvaleraldehyde
    Assay 98%

    Description

    Applications
    2-Methylvaleraldehyde is used as an intermediate to manufacture organic compounds (pharmaceuticals, odorants and flavorings)

    Solubility
    Soluble in water (4.2g/L at 25°C).

    Notes
    Keep container tightly closed. Store under inert gas. It is sensitive to air. Store in cool dry conditions in well sealed containers.

    GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit