Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-N-BOC-Amino-3-(4-tetrahydropyranyl)propionic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
500mg
Packaging:
Glass bottle
182287-51-0
C13H23NO5
273.33
MFCD02683139
NKYZORHKIYSSEL-UHFFFAOYSA-N
2-n-boc-amino-3-4-tetrahydropyranyl propionic acid,2-tert-butoxycarbonyl amino-3-tetrahydro-2h-pyran-4-yl propanoic acid,2-boc-3-tetrahydropyran-4-yl-dl-alanine,n-tert-butoxycarbonyl-3-oxan-4-ylalanine,2-tert-butoxycarbonylamino-3-tetrahydro-pyran-4-yl-propionic acid,2-tert-butoxy carbonyl amino-3-oxan-4-yl propanoic acid,2-tert-butoxycarbonyl amino-3-oxan-4-yl propanoic acid,2-2-methylpropan-2-yl oxycarbonylamino-3-oxan-4-yl propanoic acid,alpha-boc-amino tetrahydropyran-4-propanoic acid
2734406
2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(oxan-4-yl)propanoic acid
CC(C)(C)OC(=O)NC(CC1CCOCC1)C(=O)O
182287-51-0
C13H23NO5
273.33
MFCD02683139
NKYZORHKIYSSEL-UHFFFAOYSA-N
2-n-boc-amino-3-4-tetrahydropyranyl propionic acid,2-tert-butoxycarbonyl amino-3-tetrahydro-2h-pyran-4-yl propanoic acid,2-boc-3-tetrahydropyran-4-yl-dl-alanine,n-tert-butoxycarbonyl-3-oxan-4-ylalanine,2-tert-butoxycarbonylamino-3-tetrahydro-pyran-4-yl-propionic acid,2-tert-butoxy carbonyl amino-3-oxan-4-yl propanoic acid,2-tert-butoxycarbonyl amino-3-oxan-4-yl propanoic acid,2-2-methylpropan-2-yl oxycarbonylamino-3-oxan-4-yl propanoic acid,alpha-boc-amino tetrahydropyran-4-propanoic acid
2734406
2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(oxan-4-yl)propanoic acid
CC(C)(C)OC(=O)NC(CC1CCOCC1)C(=O)O
NMR 94% min.
Packaging Glass bottle
Color White
Quantity 500mg
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 273.33
Physical Form Chunks
Percent Purity 95%
Chemical Name or Material 2-N-BOC-Amino-3-(4-tetrahydropyranyl)propionic acid, 95%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning