Alfa Aesar™ 2-(N-Boc-methylamino)-5-iodo-3-methylpyridine, 95%

Manufacturer: Alfa Aesar™ H2739703

Quantity 1g

 View more versions of this product

Catalog No. AAH2739703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 942206-08-8
    Molecular Formula C12H17IN2O2
    Formula Weight 348.18
    MDL Number MFCD09037457
    Synonym tert-Butyl N-methyl-N-(5-iodo-3-methyl-2-pyridyl)carbamate
    Chemical Name or Material 2-(N-Boc-methylamino)-5-iodo-3-methylpyridine

    Specifications

    Formula Weight 348.18
    Melting Point 97° to 100°C
    Molecular Formula C12H17IN2O2
    MDL Number MFCD09037457
    CAS 942206-08-8
    Quantity 1g
    Synonym tert-Butyl N-methyl-N-(5-iodo-3-methyl-2-pyridyl)carbamate
    Chemical Name or Material 2-(N-Boc-methylamino)-5-iodo-3-methylpyridine
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit