Alfa Aesar™ 2-n-Butyl-5-ethylthiophene, 97%

Manufacturer: Alfa Aesar™ B2270814

Quantity 25g

 View more versions of this product

Catalog No. AAB2270814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 54411-06-2
    Molecular Formula C10H16S
    Formula Weight 168.3
    MDL Number MFCD00053118
    Chemical Name or Material 2-n-Butyl-5-ethylthiophene

    Specifications

    Formula Weight 168.3
    Quantity 25g
    Molecular Formula C10H16S
    MDL Number MFCD00053118
    CAS 54411-06-2
    Chemical Name or Material 2-n-Butyl-5-ethylthiophene
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit