Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-n-Butyl-5-ethylthiophene, 97%

Click to view available options
Quantity:
5g
25g
54411-06-2
C10H16S
168.298
MFCD00053118
ORAPMLHUSQJCQH-UHFFFAOYSA-N
2-n-butyl-5-ethylthiophene,thiophene, 2-butyl-5-ethyl,2-butyl-5-ethyl-thiophene,#
521504
2-butyl-5-ethylthiophene
CCCCC1=CC=C(S1)CC
54411-06-2
C10H16S
168.298
MFCD00053118
ORAPMLHUSQJCQH-UHFFFAOYSA-N
2-n-butyl-5-ethylthiophene,thiophene, 2-butyl-5-ethyl,2-butyl-5-ethyl-thiophene,#
521504
2-butyl-5-ethylthiophene
CCCCC1=CC=C(S1)CC
Quantity 5g
Formula Weight 168.3
Percent Purity 97%
Chemical Name or Material 2-n-Butyl-5-ethylthiophene