Alfa Aesar™ 2-n-Butylpropane-1,3-diol, 97%

Manufacturer: Alfa Aesar™ B2173906

Quantity 5g

 View more versions of this product

Catalog No. AAB2173906

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 2612-26-2
    Molecular Formula CH3(CH2)3CH(CH2OH)2
    Formula Weight 132.2
    MDL Number MFCD00799395
    Synonym 2-(Hydroxymethyl)hexan-1-ol
    Chemical Name or Material 2-n-Butylpropane-1,3-diol

    Specifications

    Formula Weight 132.2
    Molecular Formula CH3(CH2)3CH(CH2OH)2
    MDL Number MFCD00799395
    CAS 2612-26-2
    Synonym 2-(Hydroxymethyl)hexan-1-ol
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Quantity 5g
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 2-n-Butylpropane-1,3-diol
    Assay 97%

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit