Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-n-Butylpropane-1,3-diol, 97%

Click to view available options
Quantity:
1g
5g
2612-26-2
C7H16O2
132.203
MFCD00799395
SIMYRXPCIUXDGR-UHFFFAOYSA-N
2-butyl-1,3-propanediol,2-n-butylpropane-1,3-diol,2-n-butyl-1,3-propanediol,1,3-propanediol, 2-butyl,1-hexanol, 2-hydroxymethyl
2795259
2-butylpropane-1,3-diol
CCCCC(CO)CO
2612-26-2
C7H16O2
132.203
MFCD00799395
SIMYRXPCIUXDGR-UHFFFAOYSA-N
2-butyl-1,3-propanediol,2-n-butylpropane-1,3-diol,2-n-butyl-1,3-propanediol,1,3-propanediol, 2-butyl,1-hexanol, 2-hydroxymethyl
2795259
2-butylpropane-1,3-diol
CCCCC(CO)CO
Quantity 1g
Formula Weight 132.2
Percent Purity 97%
Chemical Name or Material 2-n-Butylpropane-1,3-diol

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger