Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(n-Hexyloxy)ethanol, 99%

Click to view available options
Quantity:
1g
5g
112-25-4
C8H18O2
146.23
MFCD00045997
UPGSWASWQBLSKZ-UHFFFAOYSA-N
2-hexyloxy ethanol,ethanol, 2-hexyloxy,2-hexyloxyethanol,ethylene glycol monohexyl ether,hexyl cellosolve,n-hexyl cellosolve,glycol monohexyl ether,cellosolve, n-hexyl,ethylene glycol n-hexyl ether
8170
2-hexoxyethanol
CCCCCCOCCO
112-25-4
C8H18O2
146.23
MFCD00045997
UPGSWASWQBLSKZ-UHFFFAOYSA-N
2-hexyloxy ethanol,ethanol, 2-hexyloxy,2-hexyloxyethanol,ethylene glycol monohexyl ether,hexyl cellosolve,n-hexyl cellosolve,glycol monohexyl ether,cellosolve, n-hexyl,ethylene glycol n-hexyl ether
8170
2-hexoxyethanol
CCCCCCOCCO
Applications
2-(n-Hexyloxy)ethanol is used as solvents in speciality printing inks and coalescing aids in surface coatings, coupling agent, rust remover, adhesives and surface cleaners.

Solubility
Soluble in alcohol and ether, water (9.46 g/L ).

Notes
Store under inert gas. Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Flash Point 94°C (201°F)
Melting Point -42°C
Quantity 1g
Boiling Point 208°C
Linear Formula CH3(CH2)5OCH2CH2OH
UN Number UN2922
Beilstein 1734691
Formula Weight 146.23
Percent Purity 99%
Chemical Name or Material 2-(n-Hexyloxy)ethanol

GHS H Statement
H314-H302-H312
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

RTECSNumber : KL2450000

TSCA : TSCA