Alfa Aesar™ 2-(n-Octyloxy)nitrobenzene

Manufacturer: Alfa Aesar™ 1574704

Quantity 2g

 View more versions of this product

Catalog No. AA1574704

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 37682-29-4
    Formula Weight 251.32
    MDL Number MFCD00014693
    Synonym o-Nitrophenyl octyl ether
    Chemical Name or Material 2-(n-Octyloxy)nitrobenzene

    Specifications

    Formula Weight 251.32
    Density 1.040g/mL
    Quantity 2g
    Flash Point >110° (230°F)
    TSCA TSCA
    Boiling Point 163° to 165°C (2mmHg)
    Physical Form Liquid
    MDL Number MFCD00014693
    CAS 37682-29-4
    Synonym o-Nitrophenyl octyl ether
    Chemical Name or Material 2-(n-Octyloxy)nitrobenzene

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit