Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2-n-Pentylthiophene, 98%
Click to view available options
Quantity:
5g
25g
Description
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. Also used as flavoring agent for food.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. Also used as flavoring agent for food.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Refractive Index | 1.4995 |
Density | 0.944 |
Boiling Point | 198°C |
Flash Point | 75°C (167°F) |
Quantity | 5g |
Beilstein | 107941 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 154.28 |
Percent Purity | 98% |
Chemical Name or Material | 2-n-Pentylthiophene |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 225-465-9
TSCA : Yes
Recommended Storage : Ambient temperatures