Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-n-Pentylthiophene, 98%

Click to view available options
Quantity:
5g
25g
4861-58-9
C9H14S
154.271
MFCD00041017
NOYVOSGVFSEKPR-UHFFFAOYSA-N
2-n-pentylthiophene,thiophene, 2-pentyl,2-n-amylthiophene,1-methylbutylthiophene,unii-l79los1zkm,l79los1zkm,2-amylthiophene,2-pentyl thiophene,2-pentyl-thiophene,acmc-209kdh
20995
2-pentylthiophene
CCCCCC1=CC=CS1
4861-58-9
C9H14S
154.271
MFCD00041017
NOYVOSGVFSEKPR-UHFFFAOYSA-N
2-n-pentylthiophene,thiophene, 2-pentyl,2-n-amylthiophene,1-methylbutylthiophene,unii-l79los1zkm,l79los1zkm,2-amylthiophene,2-pentyl thiophene,2-pentyl-thiophene,acmc-209kdh
20995
2-pentylthiophene
CCCCCC1=CC=CS1
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. Also used as flavoring agent for food.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Flash Point 75°C (167°F)
Quantity 25g
Boiling Point 198°C
Beilstein 107941
Formula Weight 154.28
Percent Purity 98%
Chemical Name or Material 2-n-Pentylthiophene

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA