Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-n-Propylfuran, 99%

Click to view available options
Quantity:
5g
25g
4229-91-8
C7H10O
110.156
MFCD00047070
CPLJMYOQYRCCBY-UHFFFAOYSA-N
furan, 2-propyl,2-n-propylfuran,2-n-propyl furan,furan, propyl,2-propyl-furan,furan, alpha-propyl,acmc-1ctud,2-propyl furan,furan, .alpha.-propyl,cpljmyoqyrccby-uhfffaoysa
77907
2-propylfuran
CCCC1=CC=CO1
4229-91-8
C7H10O
110.156
MFCD00047070
CPLJMYOQYRCCBY-UHFFFAOYSA-N
furan, 2-propyl,2-n-propylfuran,2-n-propyl furan,furan, propyl,2-propyl-furan,furan, alpha-propyl,acmc-1ctud,2-propyl furan,furan, .alpha.-propyl,cpljmyoqyrccby-uhfffaoysa
77907
2-propylfuran
CCCC1=CC=CO1
Flash Point 24°C (75°F)
Boiling Point 110°C to 111°C
Quantity 25g
UN Number UN3271
Formula Weight 110.16
Percent Purity 99%
Chemical Name or Material 2-n-Propylfuran

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.

Recommended Storage : 2° to 8°C