Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-n-Propylphenol, 97%

Manufacturer:  Alfa Aesar™ A1108814

Quantity 25g

 View more versions of this product

Catalog No. AAA1108814


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

644-35-9
136.19
UN3145
2-n-Propylphenol
C9H12O
MFCD00002251
1363932

Specifications

25g
Phenol-like
136.19
UN3145
214°C
2-n-Propylphenol
644-35-9
C9H12O
MFCD00002251
1363932
93°C (199°F)
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, LIQUID, N.O.S.

RTECSNumber : SM8600000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit