Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-n-Propylphenol 98%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
644-35-9
C9H12O
136.194
MFCD00002251
LCHYEKKJCUJAKN-UHFFFAOYSA-N
2-n-propylphenol,o-propylphenol,phenol, 2-propyl,phenol, o-propyl,1-hydroxy-2-n-propylbenzene,1-2-hydroxyphenyl propane,propylphenol,2-propyl-phenol,o-n-propylphenol,unii-333r6f6t3n
12570
2-propylphenol
CCCC1=CC=CC=C1O
644-35-9
C9H12O
136.194
MFCD00002251
LCHYEKKJCUJAKN-UHFFFAOYSA-N
2-n-propylphenol,o-propylphenol,phenol, 2-propyl,phenol, o-propyl,1-hydroxy-2-n-propylbenzene,1-2-hydroxyphenyl propane,propylphenol,2-propyl-phenol,o-n-propylphenol,unii-333r6f6t3n
12570
2-propylphenol
CCCC1=CC=CC=C1O
Density 0.9800g/mL
Quantity 25g
Refractive Index 1.5260 to 1.5290 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
Color Orange to Purple
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3CH2CH2C6H4OH
Beilstein 06, 499
Formula Weight 136.19
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2-n-Propylphenol, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 211-415-3

RTECSNumber : SM8600000

TSCA : TSCA