Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-n-Propylpyridine, 97%, Acros Organics™

Click to view available options
Quantity:
5g
25g
622-39-9
C8H11N
121.18
OIALIKXMLIAOSN-UHFFFAOYSA-N
2-n-propylpyridine,conyrine,conyrin,pyridine, 2-propyl,1-2-pyridyl propane,2-propyl-pyridine,unii-3n6ax0b7pa,2-propyl pyridine,pyridine, propyl,pyridine, 1-propyl
69320
2-propylpyridine
CCCC1=CC=CC=N1
622-39-9
C8H11N
121.18
OIALIKXMLIAOSN-UHFFFAOYSA-N
2-n-propylpyridine,conyrine,conyrin,pyridine, 2-propyl,1-2-pyridyl propane,2-propyl-pyridine,unii-3n6ax0b7pa,2-propyl pyridine,pyridine, propyl,pyridine, 1-propyl
69320
2-propylpyridine
CCCC1=CC=CC=N1
Specific Gravity 0.912
Flash Point 56°C
Density 0.9120g/mL
Infrared Spectrum Authentic
Boiling Point 169.0°C to 171.0°C
Quantity 5g
Assay Percent Range 96% min. (GC)
Formula Weight 121.18
Percent Purity 97%
Chemical Name or Material 2-n-Propylpyridine

GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Warning

EINECSNumber : 210-732-4