Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(n-Propylthio)nicotinamide, 97%

Click to view available options
Quantity:
0.25g
1g
5g
175135-26-9
C9H12N2OS
196.268
MFCD00052904
JIYBJKGFAMWNDO-UHFFFAOYSA-N
2-propylthio nicotinamide,2-n-propylthio nicotinamide,2-propylsulfanyl pyridine-3-carboxamide,2-propylthiopyridine-3-carboxamide,maybridge1_001275,2-propylthio-3-pyridinecarboxamide,3-pyridinecarboxamide,2-propylthio
2774535
2-propylsulfanylpyridine-3-carboxamide
CCCSC1=C(C=CC=N1)C(=O)N
175135-26-9
C9H12N2OS
196.268
MFCD00052904
JIYBJKGFAMWNDO-UHFFFAOYSA-N
2-propylthio nicotinamide,2-n-propylthio nicotinamide,2-propylsulfanyl pyridine-3-carboxamide,2-propylthiopyridine-3-carboxamide,maybridge1_001275,2-propylthio-3-pyridinecarboxamide,3-pyridinecarboxamide,2-propylthio
2774535
2-propylsulfanylpyridine-3-carboxamide
CCCSC1=C(C=CC=N1)C(=O)N
Quantity 1g
Melting Point 146°C to 148°C
Formula Weight 196.27
Percent Purity 97%
Chemical Name or Material 2-(n-Propylthio)nicotinamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning