Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
66-99-9
C11H8O
156.18
MFCD00004094
PJKVFARRVXDXAD-UHFFFAOYSA-N
2-naphthaldehyde,2-naphthalenecarboxaldehyde,2-formylnaphthalene,beta-naphthaldehyde,beta-formylnaphthalene,beta-naphthylaldehyde,beta-naphthylcarboxaldehyde,.beta.-naphthaldehyde,.beta.-naphthylaldehyde,.beta.-formylnaphthalene
6201
CHEBI:52368
naphthalene-2-carbaldehyde
C1=CC=C2C=C(C=CC2=C1)C=O
66-99-9
C11H8O
156.18
MFCD00004094
PJKVFARRVXDXAD-UHFFFAOYSA-N
2-naphthaldehyde,2-naphthalenecarboxaldehyde,2-formylnaphthalene,beta-naphthaldehyde,beta-formylnaphthalene,beta-naphthylaldehyde,beta-naphthylcarboxaldehyde,.beta.-naphthaldehyde,.beta.-naphthylaldehyde,.beta.-formylnaphthalene
6201
CHEBI:52368
naphthalene-2-carbaldehyde
C1=CC=C2C=C(C=CC2=C1)C=O
Packaging Glass bottle
Boiling Point 160.0°C (19.0 mmHg)
Infrared Spectrum Authentic
Color Beige or White to Yellow
Melting Point 58.0°C to 62.0°C
Flash Point >100°C
Quantity 100g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C10H7CHO
Beilstein 07, 401
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in hot water
Formula Weight 156.18
Physical Form Powder or Chunks
Percent Purity 98%
Chemical Name or Material 2-Naphthaldehyde
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning