Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthalenemethanol, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
1592-38-7
C11H10O
158.2
MFCD00004124
MFGWMAAZYZSWMY-UHFFFAOYSA-N
2-naphthalenemethanol,2-naphthylmethanol,2-hydroxymethylnaphthalene,2-naphthalene methanol,2-naphthyl methanol,naphthalen-2-yl-methanol,naphthalen-2-yl methanol,2-naphthylmethan-1-ol,2-naphthylcarbinol,2na
74128
CHEBI:27615
naphthalen-2-ylmethanol
C1=CC=C2C=C(C=CC2=C1)CO
1592-38-7
C11H10O
158.2
MFCD00004124
MFGWMAAZYZSWMY-UHFFFAOYSA-N
2-naphthalenemethanol,2-naphthylmethanol,2-hydroxymethylnaphthalene,2-naphthalene methanol,2-naphthyl methanol,naphthalen-2-yl-methanol,naphthalen-2-yl methanol,2-naphthylmethan-1-ol,2-naphthylcarbinol,2na
74128
CHEBI:27615
naphthalen-2-ylmethanol
C1=CC=C2C=C(C=CC2=C1)CO
Melting Point 79°C to 83°C
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
Color White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C10H7CH2OH
Formula Weight 158.2
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-Naphthalenemethanol
Show More Show Less