Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthalenethiol, 98+%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
91-60-1
C10H8S
160.2
MFCD00004086
RFCQDOVPMUSZMN-UHFFFAOYSA-N
2-naphthalenethiol,2-mercaptonaphthalene,2-thionaphthol,thionaphthol,2-naphthyl mercaptan,vulcamel tbn,2-naphthyl thiol,renacit 1,rpa no. 2,rpa 2
7058
naphthalene-2-thiol
C1=CC=C2C=C(C=CC2=C1)S
91-60-1
C10H8S
160.2
MFCD00004086
RFCQDOVPMUSZMN-UHFFFAOYSA-N
2-naphthalenethiol,2-mercaptonaphthalene,2-thionaphthol,thionaphthol,2-naphthyl mercaptan,vulcamel tbn,2-naphthyl thiol,renacit 1,rpa no. 2,rpa 2
7058
naphthalene-2-thiol
C1=CC=C2C=C(C=CC2=C1)S
Boiling Point 286°C
Melting Point 79°C to 83°C
Packaging Glass bottle
Quantity 25g
Color Cream to White
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98% min. (GC)
Merck Index 15, 6464
Solubility Information Solubility in water: .. Other solubilities: soluble in organic solvents
Formula Weight 160.2
Physical Form Powder
Percent Purity 98+%
Chemical Name or Material 2-Naphthalenethiol, 99%
Show More Show Less

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.

GHS Signal Word: Warning

EINECSNumber : 202-082-5

RTECSNumber : QK3930000

TSCA : TSCA