Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthoic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
93-09-4
C11H8O2
172.18
MFCD00004101
UOBYKYZJUGYBDK-UHFFFAOYSA-N
2-naphthoic acid,2-naphthalenecarboxylic acid,isonaphthoic acid,2-carboxynaphthalene,2-maythic acid,beta-naphthoic acid,ne-2-carboxylic acid,unii-qlg01v0w2l,naphthalene-beta-carboxylic acid,.beta.-naphthoic acid
7123
CHEBI:36106
naphthalene-2-carboxylic acid
C1=CC=C2C=C(C=CC2=C1)C(=O)O
93-09-4
C11H8O2
172.18
MFCD00004101
UOBYKYZJUGYBDK-UHFFFAOYSA-N
2-naphthoic acid,2-naphthalenecarboxylic acid,isonaphthoic acid,2-carboxynaphthalene,2-maythic acid,beta-naphthoic acid,ne-2-carboxylic acid,unii-qlg01v0w2l,naphthalene-beta-carboxylic acid,.beta.-naphthoic acid
7123
CHEBI:36106
naphthalene-2-carboxylic acid
C1=CC=C2C=C(C=CC2=C1)C(=O)O
Color White to Beige
Boiling Point >300°C
Flash Point 205°C
Density 205g/mL
Melting Point 183°C to 187°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (HPLC)
Linear Formula C10H7CO2H
Beilstein 09, 656
Merck Index 15, 6467
Solubility Information Solubility in water: <0.5g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 172.18
Physical Form Powder
Percent Purity 99%
Chemical Name or Material 2-Naphthoic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 202-217-8