Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthol, 99+%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
1kg
2.5kg
10kg
Packaging:
Glass bottle
Plastic drum
135-19-3
C10H8O
144.17
MFCD00004067
JWAZRIHNYRIHIV-UHFFFAOYSA-N
2-naphthol,2-naphthalenol,beta-naphthol,2-hydroxynaphthalene,betanaphthol,isonaphthol,developer bn,naphthol b,azogen developer a,beta-napthol
8663
CHEBI:10432
naphthalen-2-ol
C1=CC=C2C=C(C=CC2=C1)O
135-19-3
C10H8O
144.17
MFCD00004067
JWAZRIHNYRIHIV-UHFFFAOYSA-N
2-naphthol,2-naphthalenol,beta-naphthol,2-hydroxynaphthalene,betanaphthol,isonaphthol,developer bn,naphthol b,azogen developer a,beta-napthol
8663
CHEBI:10432
naphthalen-2-ol
C1=CC=C2C=C(C=CC2=C1)O
Flash Point 153°C
Packaging Plastic drum
Melting Point 120.0°C to 124.0°C
Boiling Point 285.0°C to 286.0°C
Color White
Quantity 10kg
Infrared Spectrum Authentic
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99% min. (GC)
Linear Formula C10H7OH
Beilstein 06,627
Merck Index 15,6469
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: 1 g/80mL boiling water, 1 g/0.8mL alcohol, 1 g/17mL chloroform, 1 g/1.3mL ether, soluble in ethanol, ether, chloroform, glycerol
Formula Weight 144.17
Physical Form Powder, Crystals or Granules
Percent Purity 99+%
Chemical Name or Material 2-Naphthol, 99+%
Show More Show Less

GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Very toxic to aquatic life.

GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 205-182-7

RTECSNumber : QL2975000

TSCA : TSCA