Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthol, Purified, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
135-19-3
135-19-3
Packaging Fiber Drum
Quantity 12 kg
CAS Max % 1
Grade Purified

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.