Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthoxyacetic Acid 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
120-23-0
C12H10O3
202.209
MFCD00004066
RZCJYMOBWVJQGV-UHFFFAOYSA-N
2-naphthoxyacetic acid,2-naphthyloxy acetic acid,bnoa,2-naphthyloxyacetic acid,2-naphthalen-2-yloxy acetic acid,noxa,betoxon,2-noxa,naphthoxyacetic acid,betapal
8422
CHEBI:50397
2-naphthalen-2-yloxyacetic acid
C1=CC=C2C=C(C=CC2=C1)OCC(=O)O
120-23-0
C12H10O3
202.209
MFCD00004066
RZCJYMOBWVJQGV-UHFFFAOYSA-N
2-naphthoxyacetic acid,2-naphthyloxy acetic acid,bnoa,2-naphthyloxyacetic acid,2-naphthalen-2-yloxy acetic acid,noxa,betoxon,2-noxa,naphthoxyacetic acid,betapal
8422
CHEBI:50397
2-naphthalen-2-yloxyacetic acid
C1=CC=C2C=C(C=CC2=C1)OCC(=O)O
Melting Point 154°C to 158°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Loss on Drying 0.5% max.
Color Blue to Gray-Green
Quantity 500g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C10H7OCH2CO2H
Beilstein 06, 645
Merck Index 15, 6482
Formula Weight 202.21
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-Naphthoxyacetic acid, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 204-380-0

RTECSNumber : AI9659000

TSCA : TSCA