Alfa Aesar™ 2-Naphthyl isocyanate, 97%

Manufacturer: Alfa Aesar™ H5107603

Quantity 1g

 View more versions of this product

Catalog No. AAH5107603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2243-54-1
    Molecular Formula C11H7NO
    Formula Weight 169.18
    MDL Number MFCD00014319
    UN Number UN2811
    Chemical Name or Material 2-Naphthyl isocyanate

    Specifications

    Formula Weight 169.18
    Molecular Formula C11H7NO
    Melting Point 53° to 56°C
    MDL Number MFCD00014319
    CAS 2243-54-1
    UN Number UN2811
    Boiling Point 83°C (0.2mmHg)
    Quantity 1g
    Odor Pungent
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H334-H315-H319-H335
    Toxic if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P260-P280h-P305+P351+P338-P309-P310
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material 2-Naphthyl isocyanate
    Assay 97%

    Description

    Applications
    2-Naphthyl isocyanate is used as pharmaceutical intermediate.

    Solubility
    It hydrolyzes in water.

    Notes
    Moisture sensitive. Incompatible with oxidizing agents, alcohols and amines.

    GHS H Statement
    H331-H334-H315-H319-H335
    Toxic if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P260-P280h-P305+P351+P338-P309-P310
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit