Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthylacetonitrile, 90-95%, Tech., ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
7498-57-9
C12H9N
167.211
MFCD00004122
LPCWDVLDJVZIHA-UHFFFAOYSA-N
2-naphthylacetonitrile,2-naphthalen-2-yl acetonitrile,2-naphthaleneacetonitrile,2-naphthyl acetonitrile,naphthalene-2-acetonitrile,2-2-naphthyl acetonitrile,.beta.-naphthyleneacetonitrile,2-naphtylacetonitrile,2-napthylacetonitrile,2-naphthylmethylcyanide
82008
2-naphthalen-2-ylacetonitrile
C1=CC=C2C=C(C=CC2=C1)CC#N
7498-57-9
C12H9N
167.211
MFCD00004122
LPCWDVLDJVZIHA-UHFFFAOYSA-N
2-naphthylacetonitrile,2-naphthalen-2-yl acetonitrile,2-naphthaleneacetonitrile,2-naphthyl acetonitrile,naphthalene-2-acetonitrile,2-2-naphthyl acetonitrile,.beta.-naphthyleneacetonitrile,2-naphtylacetonitrile,2-napthylacetonitrile,2-naphthylmethylcyanide
82008
2-naphthalen-2-ylacetonitrile
C1=CC=C2C=C(C=CC2=C1)CC#N
Quantity 5g
Packaging Glass bottle
Density 1.0900g/mL
CAS Min % 90.0
CAS Max % 100.0
Assay Percent Range 90-95%
Linear Formula C10H7CH2CN
Beilstein 09, 659
Formula Weight 167.21
Grade Technical
Chemical Name or Material 2-Naphthylacetonitrile, tech., 90-95%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning