Learn More
BioVision IncSupplier Diversity Partner 2-NBDG Glucose Uptake Assay Kit (Cell-Based): 2-NBDG Glucose Uptake Assay Kit (Cell-Based): Easy-to-use, Non-radioactive method to image and accurately measure glucose uptake in cultured cells via FACS and microscopy. 50 assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K68250
Glucose is a ubiquitous energy source in most organisms and plays a pivotal role in cellular metabolisms and homeostasis. Cancer cells exhibit increased glucose uptake to support their high proliferation rate. 2-NBDG (2-deoxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl) amino]-D-glucose) is a fluorescent deoxyglucose analog that can be taken up by cells through glucose transporters. However, 2-NBDG cannot be fully utilized in glycolysis because of its modification and thus accumulates inside the cells. Fluorescence generated by this fluorescent glucose analog is proportional to glucose uptake by the cells and can be used to measure glucose uptake using fluorescent microscopy and flow cytometry. To validate the assay, the kit includes phloretin, a natural phenol that inhibits glucose uptake. This easy to use non-radioactive kit allows imaging and accurate measurement of glucose uptake in cultured cells in response to insulin, growth factors etc.
Catalog No. 50-111-9837