Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitro-2-methyl-1,3-propanediol 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
77-49-6
C4H9NO4
135.12
MFCD00024812
LOTYADDQWWVBDJ-UHFFFAOYSA-N
2-methyl-2-nitro-1,3-propanediol,nmpd,2-nitro-2-methyl-1,3-propanediol,nitroisobutylglycol,1,3-propanediol, 2-methyl-2-nitro,2-methyl-2-nitropropanediol,1,1-dimethylol-1-nitroethane,caswell no. 573n,epa pesticide chemical code 573900,acmc-1basy
6480
2-methyl-2-nitropropane-1,3-diol
CC(CO)(CO)[N+](=O)[O-]
77-49-6
C4H9NO4
135.12
MFCD00024812
LOTYADDQWWVBDJ-UHFFFAOYSA-N
2-methyl-2-nitro-1,3-propanediol,nmpd,2-nitro-2-methyl-1,3-propanediol,nitroisobutylglycol,1,3-propanediol, 2-methyl-2-nitro,2-methyl-2-nitropropanediol,1,1-dimethylol-1-nitroethane,caswell no. 573n,epa pesticide chemical code 573900,acmc-1basy
6480
2-methyl-2-nitropropane-1,3-diol
CC(CO)(CO)[N+](=O)[O-]
Melting Point 147°C to 149°C
Quantity 5g
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Solubility Information Solubility in water: 80g/100mL (20 c)
Formula Weight 135.12
Chemical Name or Material 2-Nitro-2-methyl-1, 3-propanediol, 99%

EINECSNumber : 201-031-4

RTECSNumber : TY8925000

TSCA : TSCA