Alfa Aesar™ 2-Nitro-4-(trifluoromethyl)anisole, 98%

Manufacturer: Alfa Aesar™ A1184722

Quantity 100g

 View more versions of this product

Catalog No. AAA1184722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 394-25-2
    Molecular Formula C8H6F3NO3
    Formula Weight 221.14
    MDL Number MFCD00007099
    UN Number UN1759
    Beilstein 2127954
    Synonym 4-Methoxy-3-nitrobenzotrifluoride
    Chemical Name or Material 2-Nitro-4- (trifluoromethyl)anisole

    Specifications

    Formula Weight 221.14
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 394-25-2
    MDL Number MFCD00007099
    Boiling Point 96° to 98°C (0.6mmHg)
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
    UN Number UN1759
    Flash Point >110°C (230°F)
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Beilstein 2127954
    Quantity 100g
    Molecular Formula C8H6F3NO3
    Melting Point 44° to 49°C
    Synonym 4-Methoxy-3-nitrobenzotrifluoride
    Chemical Name or Material 2-Nitro-4- (trifluoromethyl)anisole
    Assay 98%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit