Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitro-4-(trifluoromethyl)benzoic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
320-94-5
C8H4F3NO4
235.11
MYSAXQPTXWKDPQ-UHFFFAOYSA-N
2-nitro-4-trifluoromethyl benzoic acid,2-nitro-4-trifluoromethylbenzoic acid,ntfa,4-trifluoromethyl-2-nitrobenzoic acid,benzoic acid, 2-nitro-4-trifluoromethyl,unii-k9m35sv4p3,2-nitro-4-trifluoromethyl-benzoic acid,pubchem4539,acmc-1ahai,2ntf-boa
642096
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(=O)O
320-94-5
C8H4F3NO4
235.11
MYSAXQPTXWKDPQ-UHFFFAOYSA-N
2-nitro-4-trifluoromethyl benzoic acid,2-nitro-4-trifluoromethylbenzoic acid,ntfa,4-trifluoromethyl-2-nitrobenzoic acid,benzoic acid, 2-nitro-4-trifluoromethyl,unii-k9m35sv4p3,2-nitro-4-trifluoromethyl-benzoic acid,pubchem4539,acmc-1ahai,2ntf-boa
642096
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(=O)O
Packaging Glass Bottle
Infrared Spectrum Authentic
Color Yellow
Quantity 1g
Melting Point 137.0°C to 140.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 235.11
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-Nitro-4-(trifluoromethyl)benzoic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning