Alfa Aesar™ 2-Nitro-9-fluorenone, 99%

Manufacturer: Alfa Aesar™ H5569903

Quantity 1g

 View more versions of this product

Catalog No. AAH5569903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3096-52-4
    Formula Weight 225.2
    MDL Number MFCD00001152
    Chemical Name or Material 2-Nitro-9-fluorenone

    Specifications

    Formula Weight 225.2
    Melting Point 222° to 223°C
    Physical Form Solid
    CAS 3096-52-4
    MDL Number MFCD00001152
    RTECS Number LL9091000
    Quantity 1g
    Chemical Name or Material 2-Nitro-9-fluorenone
    Assay 99%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit