Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Nitro-p-phenylenediamine, 95%

Click to view available options
Quantity:
25g
100g
500g
5307-14-2
C6H7N3O2
153.141
MFCD00007903
HVHNMNGARPCGGD-UHFFFAOYSA-N
2-nitro-1,4-phenylenediamine,1,4-diamino-2-nitrobenzene,2-nitro-p-phenylenediamine,2,5-diaminonitrobenzene,1,4-benzenediamine, 2-nitro,durafur brown,2-nitro-1,4-benzenediamine,o-nitro-p-phenylenediamine,4-amino-2-nitroaniline,fouramine 2r
4338370
CHEBI:76394
2-nitrobenzene-1,4-diamine
C1=CC(=C(C=C1N)[N+](=O)[O-])N
5307-14-2
C6H7N3O2
153.141
MFCD00007903
HVHNMNGARPCGGD-UHFFFAOYSA-N
2-nitro-1,4-phenylenediamine,1,4-diamino-2-nitrobenzene,2-nitro-p-phenylenediamine,2,5-diaminonitrobenzene,1,4-benzenediamine, 2-nitro,durafur brown,2-nitro-1,4-benzenediamine,o-nitro-p-phenylenediamine,4-amino-2-nitroaniline,fouramine 2r
4338370
CHEBI:76394
2-nitrobenzene-1,4-diamine
C1=CC(=C(C=C1N)[N+](=O)[O-])N
Melting Point 134°C to 137°C
Quantity 25g
Formula Weight 153.14
Percent Purity 95%
Chemical Name or Material 2-Nitro-p-phenylenediamine

GHS H Statement
H341-H351
Suspected of causing genetic defects.
Suspected of causing cancer.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : ST3000000

TSCA : TSCA