Alfa Aesar™ 2-Nitro-p-phenylenediamine, 95%

Manufacturer: Alfa Aesar™ B2347736

Quantity 500g

 View more versions of this product

Catalog No. AAB2347736

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5307-14-2
    Molecular Formula C6H7N3O2
    Formula Weight 153.14
    MDL Number MFCD00007903
    Synonym 2,5-Diaminonitrobenzene
    Chemical Name or Material 2-Nitro-p-phenylenediamine

    Specifications

    Formula Weight 153.14
    TSCA TSCA
    MDL Number MFCD00007903
    Synonym 2,5-Diaminonitrobenzene
    Health Hazard 2 GHS H Statement
    H341-H351
    Suspected of causing genetic defects.
    Suspected of causing cancer.
    RTECS Number ST3000000
    Melting Point 134° to 137°C
    Quantity 500g
    Molecular Formula C6H7N3O2
    CAS 5307-14-2
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 2-Nitro-p-phenylenediamine
    Assay 95%

    GHS H Statement
    H341-H351
    Suspected of causing genetic defects.
    Suspected of causing cancer.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit