Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitroaniline, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
5kg
Packaging:
Glass bottle
Plastic drum
88-74-4
C6H6N2O2
138.13
MFCD00007687
DPJCXCZTLWNFOH-UHFFFAOYSA-N
o-nitroaniline,benzenamine, 2-nitro,o-nitraniline,1-amino-2-nitrobenzene,2-nitrobenzenamine,aniline, o-nitro,devol orange b,orange grs salt,o-aminonitrobenzene,2-aminonitrobenzene
6946
2-nitroaniline
C1=CC=C(C(=C1)N)[N+](=O)[O-]
88-74-4
C6H6N2O2
138.13
MFCD00007687
DPJCXCZTLWNFOH-UHFFFAOYSA-N
o-nitroaniline,benzenamine, 2-nitro,o-nitraniline,1-amino-2-nitrobenzene,2-nitrobenzenamine,aniline, o-nitro,devol orange b,orange grs salt,o-aminonitrobenzene,2-aminonitrobenzene
6946
2-nitroaniline
C1=CC=C(C(=C1)N)[N+](=O)[O-]
Indicator
Quantity 5g
Melting Point 70.0°C to 74.0°C
Flash Point 168°C
Infrared Spectrum Authentic
Color Orange to Brown
Boiling Point 284.0°C
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula O2NC6H4NH2
Beilstein 12,687
Merck Index 15,6669
Solubility Information Solubility in water: 1.1g/L in water (20°C). Other solubilities: soluble in alcohol,chloroform and ether
Formula Weight 138.13
Physical Form Crystals or Flakes
Percent Purity 98%
Chemical Name or Material 2-Nitroaniline, 98%
Show More Show Less

GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Harmful to aquatic life with long lasting effects.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-855-4

RTECSNumber : BY6650000

TSCA : TSCA