Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrobenzaldehyde, 99+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Plastic bottle
552-89-6
C7H5NO3
151.12
CMWKITSNTDAEDT-UHFFFAOYSA-N
o-nitrobenzaldehyde,benzaldehyde, 2-nitro,nitrobenzaldehyde,benzaldehyde, o-nitro,2-nitro-benzaldehyde,2-formylnitrobenzene,ccris 2322,unii-48b18q9b8e,benzaldehyde, nitro,ortho-nitrobenzaldehyde
11101
CHEBI:66927
2-nitrobenzaldehyde
C1=CC=C(C(=C1)C=O)[N+](=O)[O-]
552-89-6
C7H5NO3
151.12
CMWKITSNTDAEDT-UHFFFAOYSA-N
o-nitrobenzaldehyde,benzaldehyde, 2-nitro,nitrobenzaldehyde,benzaldehyde, o-nitro,2-nitro-benzaldehyde,2-formylnitrobenzene,ccris 2322,unii-48b18q9b8e,benzaldehyde, nitro,ortho-nitrobenzaldehyde
11101
CHEBI:66927
2-nitrobenzaldehyde
C1=CC=C(C(=C1)C=O)[N+](=O)[O-]
Boiling Point 153.0°C (23.0 mmHg)
Color Yellow
Infrared Spectrum Authentic
Flash Point >110°C
Packaging Plastic bottle
Melting Point 41.0°C to 44.0°C
Quantity 100g
Assay Percent Range 99+%
Linear Formula O2NC6H4CHO
Beilstein 07,243
Merck Index 14,6587
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in alcohol,benzene and ether
Formula Weight 151.12
Physical Form Crystalline Powder, Needles and/or Chunks
Percent Purity ≥99%
Chemical Name or Material 2-Nitrobenzaldehyde
Show More Show Less

GHS H Statement
Harmful to aquatic life with long lasting effects. Harmful if swallowed. Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation.

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

Warning