Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrobenzamide, 98%, Acros Organics

Click to view available options
Quantity:
50g
610-15-1
C7H6N2O3
166.136
MFCD00007976
KLGQWSOYKYFBTR-UHFFFAOYSA-N
o-nitrobenzamide,benzamide, o-nitro,benzamide, 2-nitro,2-carbamoylnitrobenzene,nitrobenzamide,nitro-benzamide,2-nitro-benzamide,benzamide,2-nitro,benzmide, 2-nitro,wln: zvr bnw
11876
2-nitrobenzamide
C1=CC=C(C(=C1)C(=O)N)[N+](=O)[O-]
610-15-1
C7H6N2O3
166.136
MFCD00007976
KLGQWSOYKYFBTR-UHFFFAOYSA-N
o-nitrobenzamide,benzamide, o-nitro,benzamide, 2-nitro,2-carbamoylnitrobenzene,nitrobenzamide,nitro-benzamide,2-nitro-benzamide,benzamide,2-nitro,benzmide, 2-nitro,wln: zvr bnw
11876
2-nitrobenzamide
C1=CC=C(C(=C1)C(=O)N)[N+](=O)[O-]
Color Beige
Infrared Spectrum Authentic
Melting Point 174°C to 178°C
Quantity 50g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula O2NC6H4CONH2
Beilstein 09, 373
Formula Weight 166.14
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-Nitrobenzamide, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 210-208-5

RTECSNumber : CV5600000

TSCA : TSCA