Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Nitrobenzenesulfenyl chloride, 98%

Click to view available options
Quantity:
10g
50g
7669-54-7
C6H4ClNO2S
189.613
MFCD00007128
NTNKNFHIAFDCSJ-UHFFFAOYSA-N
2-nitrobenzenesulfenyl chloride,2-nitrophenylsulfenyl chloride,benzenesulfenyl chloride, 2-nitro,o-nitrobenzenesulfenyl chloride,o-nitrophenylsulfenyl chloride,2-nitrobenzenesulfenylchloride,o-nitrophenylsulphenyl chloride,o-nitrobenzenesulphenyl chloride,2-nitrophenyl thiohypochlorite,2-nitrophenyl sulfanyl chlorane
24319
(2-nitrophenyl) thiohypochlorite
C1=CC=C(C(=C1)[N+](=O)[O-])SCl
7669-54-7
C6H4ClNO2S
189.613
MFCD00007128
NTNKNFHIAFDCSJ-UHFFFAOYSA-N
2-nitrobenzenesulfenyl chloride,2-nitrophenylsulfenyl chloride,benzenesulfenyl chloride, 2-nitro,o-nitrobenzenesulfenyl chloride,o-nitrophenylsulfenyl chloride,2-nitrobenzenesulfenylchloride,o-nitrophenylsulphenyl chloride,o-nitrobenzenesulphenyl chloride,2-nitrophenyl thiohypochlorite,2-nitrophenyl sulfanyl chlorane
24319
(2-nitrophenyl) thiohypochlorite
C1=CC=C(C(=C1)[N+](=O)[O-])SCl
Applications
2-Nitrobenzenesulfenyl chloride is used as a reagent for N-protection as N-2-nitrophenylsulfenyl (Nps) derivatives, in amino acids and peptides and nucleosides.

Solubility
Reacts with water.

Notes
Store under dry inert gas. Store away from moisture, strong bases and oxidizing agents. Protect from humidity and water.
Melting Point 72°C to 75°C
Quantity 50g
UN Number UN3261
Beilstein 473741
Formula Weight 189.62
Percent Purity 98%
Chemical Name or Material 2-Nitrobenzenesulfenyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA

Recommended Storage : 2° to 8°C