Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrobenzenesulfonamide, 96%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
5455-59-4
C6H6N2O4S
202.184
MFCD00009807
GNDKYAWHEKZHPJ-UHFFFAOYSA-N
2-nitrobenzene-1-sulfonamide,2-nitrobenzenesulphonamide,2-nitro-benzenesulfonamide,nitrobenzenesulfonamide,2-nitrophenylsulfonamide,2-sulphamoylnitrobenzene,acmc-209lhg,2-nitrobenzene-sulfonamide,2-nitro benzene sulphonamide,ksc490i0n
138510
2-nitrobenzenesulfonamide
C1=CC=C(C(=C1)[N+](=O)[O-])S(=O)(=O)N
5455-59-4
C6H6N2O4S
202.184
MFCD00009807
GNDKYAWHEKZHPJ-UHFFFAOYSA-N
2-nitrobenzene-1-sulfonamide,2-nitrobenzenesulphonamide,2-nitro-benzenesulfonamide,nitrobenzenesulfonamide,2-nitrophenylsulfonamide,2-sulphamoylnitrobenzene,acmc-209lhg,2-nitrobenzene-sulfonamide,2-nitro benzene sulphonamide,ksc490i0n
138510
2-nitrobenzenesulfonamide
C1=CC=C(C(=C1)[N+](=O)[O-])S(=O)(=O)N
Color Yellow to Brown
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 189°C to 194°C
Quantity 25g
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula O2NC6H4SO2NH2
Beilstein 11, 68
Formula Weight 202.18
Physical Form Crystalline Powder
Percent Purity ≥95%
Chemical Name or Material 2-Nitrobenzenesulfonamide
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.