Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrobenzonitrile, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
100g
Packaging:
Glass bottle
612-24-8
C7H4N2O2
148.12
SWBDKCMOLSUXRH-UHFFFAOYSA-N
o-nitrobenzonitrile,o-cyanonitrobenzene,benzonitrile, o-nitro,benzonitrile, 2-nitro,2-cyanonitrobenzene,nitrobenzonitrile,unii-drc6u29fci,ccris 2326,1-nitro-2-cyanobenzene,2-nitrobenzenecarbonitrile
11922
2-nitrobenzonitrile
C1=CC=C(C(=C1)C#N)[N+](=O)[O-]
612-24-8
C7H4N2O2
148.12
SWBDKCMOLSUXRH-UHFFFAOYSA-N
o-nitrobenzonitrile,o-cyanonitrobenzene,benzonitrile, o-nitro,benzonitrile, 2-nitro,2-cyanonitrobenzene,nitrobenzonitrile,unii-drc6u29fci,ccris 2326,1-nitro-2-cyanobenzene,2-nitrobenzenecarbonitrile
11922
2-nitrobenzonitrile
C1=CC=C(C(=C1)C#N)[N+](=O)[O-]
Packaging Glass bottle
Boiling Point 165.0°C (16.0 mmHg)
Infrared Spectrum Authentic
Quantity 10g
Color Yellow
Melting Point 110.8°C to 111.8°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula O2NC6H4CN
Beilstein 09,374
Solubility Information Solubility in water: insoluble
Formula Weight 148.12
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-Nitrobenzonitrile
Show More Show Less

GHS H Statement:
Fatal if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger