Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrobenzyl Bromide 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
3958-60-9
C7H6BrNO2
216.03
MFCD00007184
HXBMIQJOSHZCFX-UHFFFAOYSA-N
2-nitrobenzyl bromide,1-bromomethyl-2-nitrobenzene,o-nitrobenzyl bromide,alpha-bromo-2-nitrotoluene,2-bromomethyl nitrobenzene,1-bromomethyl-2-nitro-benzene,benzene, 1-bromomethyl-2-nitro,ccris 7966,2-nitro benzyl bromide,2-nitro-benzyl bromide
77569
1-(bromomethyl)-2-nitrobenzene
C1=CC=C(C(=C1)CBr)[N+](=O)[O-]
3958-60-9
C7H6BrNO2
216.03
MFCD00007184
HXBMIQJOSHZCFX-UHFFFAOYSA-N
2-nitrobenzyl bromide,1-bromomethyl-2-nitrobenzene,o-nitrobenzyl bromide,alpha-bromo-2-nitrotoluene,2-bromomethyl nitrobenzene,1-bromomethyl-2-nitro-benzene,benzene, 1-bromomethyl-2-nitro,ccris 7966,2-nitro benzyl bromide,2-nitro-benzyl bromide
77569
1-(bromomethyl)-2-nitrobenzene
C1=CC=C(C(=C1)CBr)[N+](=O)[O-]
Flash Point >110°C
Melting Point 44°C to 48°C
Color Yellow to Brown
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula O2NC6H4CH2Br
Beilstein 05, I, 164
Solubility Information Solubility in water: insoluble
Formula Weight 216.03
Physical Form Crystals or Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-Nitrobenzyl bromide, 97%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 223-558-9

TSCA : TSCA