Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrobenzyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
612-23-7
C7H6ClNO2
171.58
MFCD00007185
BXCBUWKTXLWPSB-UHFFFAOYSA-N
2-nitrobenzyl chloride,1-chloromethyl-2-nitrobenzene,o-nitrobenzyl chloride,alpha-chloro-2-nitrotoluene,benzene, 1-chloromethyl-2-nitro,2-nitrobenzylchloride,unii-hb8u484npm,alpha-chloro-o-nitrotoluene,ccris 2323,toluene, alpha-chloro-o-nitro
11921
1-(chloromethyl)-2-nitrobenzene
C1=CC=C(C(=C1)CCl)[N+](=O)[O-]
612-23-7
C7H6ClNO2
171.58
MFCD00007185
BXCBUWKTXLWPSB-UHFFFAOYSA-N
2-nitrobenzyl chloride,1-chloromethyl-2-nitrobenzene,o-nitrobenzyl chloride,alpha-chloro-2-nitrotoluene,benzene, 1-chloromethyl-2-nitro,2-nitrobenzylchloride,unii-hb8u484npm,alpha-chloro-o-nitrotoluene,ccris 2323,toluene, alpha-chloro-o-nitro
11921
1-(chloromethyl)-2-nitrobenzene
C1=CC=C(C(=C1)CCl)[N+](=O)[O-]
Boiling Point 127°C to 133°C (10.0mmHg)
Flash Point >110°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color Tan to Yellow
Quantity 50g
Melting Point 44°C to 49°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula O2NC6H4CH2Cl
Beilstein 05, 327
Solubility Information Solubility in water: insoluble in water
Formula Weight 171.58
Physical Form Crystals, Crystalline Powder or Chunks
Percent Purity ≥97.5%
Chemical Name or Material 2-Nitrobenzyl chloride, 98%
Show More Show Less

GHS P Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger

EINECSNumber : 210-300-5

RTECSNumber : XS9092000

TSCA : TSCA