Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Nitrocinnamaldehyde, predominantly trans, 98%

Manufacturer:  Alfa Aesar™ A1030814

Quantity 25g

 View more versions of this product

Catalog No. AAA1030814


  Check Availability
Add to cart

Specifications

Description

Applications
It finds its uses as a actinometer for the UV-A range of photostability testing of pharmaceuticals. 2-Nitrocinnamaldehyde can be oxidized to 2-nitrocinnamic acid which can be used in the Baeyer-Emmerling indole synthesis to produce indole and substituted indoles.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Description & Specifications

Chemical Identifiers

1466-88-2
177.16
908376
C9H7NO3
MFCD00007188
2-Nitrocinnamaldehyde, predominantly trans

Specifications

Characteristic
25g
C9H7NO3
MFCD00007188
2-Nitrocinnamaldehyde, predominantly trans
125° to 129°C
1466-88-2
177.16
908376
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit