Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Nitroethanol, tech. 80%

Click to view available options
Quantity:
5g
25g
100g
625-48-9
C2H5NO3
91.066
MFCD00007405
KIPMDPDAFINLIV-UHFFFAOYSA-N
ethanol, 2-nitro,2-nitroethan-1-ol,beta-nitroalcohol,beta-nitroethanol,2-nitro-ethanol,.beta.-nitroethanol,ccris 6076,2 nitroethanol,pubchem14208,2-nitroethanol
12252
2-nitroethanol
C(CO)[N+](=O)[O-]
625-48-9
C2H5NO3
91.066
MFCD00007405
KIPMDPDAFINLIV-UHFFFAOYSA-N
ethanol, 2-nitro,2-nitroethan-1-ol,beta-nitroalcohol,beta-nitroethanol,2-nitro-ethanol,.beta.-nitroethanol,ccris 6076,2 nitroethanol,pubchem14208,2-nitroethanol
12252
2-nitroethanol
C(CO)[N+](=O)[O-]
Refractive Index 1.4451
Density 1.27
Quantity 5g
Melting Point -80°C
Flash Point >110°C (230°F)
Boiling Point 194°C
Beilstein 1633753
Formula Weight 91.07
Percent Purity 80%
Grade Technical
Chemical Name or Material 2-Nitroethanol
Show More Show Less

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 210-895-1

RTECSNumber : KL9370000

TSCA : No

Recommended Storage : Ambient temperatures