Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Nitromethylene)thiazolidine, 97%

Click to view available options
Quantity:
1g
5g
66357-40-2
C4H6N2O2S
146.164
MFCD04039774
IDEZECZCLMOIJN-ONEGZZNKSA-N
2-nitromethylenethiazolidine,2-nitromethylene thiazolidine,2e-2-nitromethylidene-1,3-thiazolidine,thiazolidine, 2-nitromethylene,2-e-nitromethylene thiazolidine,2e-2-nitromethylene thiazolidine
6455311
(2E)-2-(nitromethylidene)-1,3-thiazolidine
C1CSC(=C[N+](=O)[O-])N1
66357-40-2
C4H6N2O2S
146.164
MFCD04039774
IDEZECZCLMOIJN-ONEGZZNKSA-N
2-nitromethylenethiazolidine,2-nitromethylene thiazolidine,2e-2-nitromethylidene-1,3-thiazolidine,thiazolidine, 2-nitromethylene,2-e-nitromethylene thiazolidine,2e-2-nitromethylene thiazolidine
6455311
(2E)-2-(nitromethylidene)-1,3-thiazolidine
C1CSC(=C[N+](=O)[O-])N1
Quantity 1g
Melting Point 141°C to 143°C
Formula Weight 146.16
Percent Purity 97%
Chemical Name or Material 2-(Nitromethylene)thiazolidine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning