Alfa Aesar™ 2-Nitrophenol, 98%

Manufacturer: Alfa Aesar™ A123330I

Quantity 5000g

 View more versions of this product

Catalog No. AAA123330I

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 88-75-5
    Molecular Formula C6H5NO3
    Formula Weight 139.11
    MDL Number MFCD00011688
    UN Number UN1663
    Beilstein 775403
    Merck Index 14,6620
    Chemical Name or Material 2-Nitrophenol

    Specifications

    Formula Weight 139.11
    MDL Number MFCD00011688
    CAS 88-75-5
    Health Hazard 3 GHS P Statement
    P280h-P273-P302+P352
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF ON SKIN: Wash with plenty of soap and water.
    RTECS Number SM2100000
    Melting Point 44° to 48°C
    Quantity 5000g
    UN Number UN1663
    Beilstein 775403
    Merck Index 14,6620
    Health Hazard 1 Warning
    Flash Point 108°C (226°F)
    TSCA TSCA
    Molecular Formula C6H5NO3
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROPHENOLS
    Boiling Point 214° to 216°C
    Health Hazard 2 GHS H Statement
    H373-H302
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Odor Phenol-like
    Chemical Name or Material 2-Nitrophenol
    Assay 98%

    Description

    Applications
    2-Nitrophenol is a building block used as an acid-base indicator and corrosion inhibitor for aluminum-copper alloy. It is also used to make dyes, paint coloring, rubber chemicals, and substances that kill molds.

    Solubility
    Soluble in water (2.1 g/L at 20°C), ethanol, chloroform, alcohol, and acetone.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.

    GHS H Statement
    H373-H302
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.

    GHS P Statement
    P280h-P273-P302+P352
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF ON SKIN: Wash with plenty of soap and water.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit